missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TREM1 Protein

A cDNA sequence encoding the TREM1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13808-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984769

  • 3401.86 € / 1mg

Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TREM1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TREM1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMATKLTE EKYELKEGQT LDVKCDYTLE KFASSQKAWQ IIRDGEMPKT LACTERPSKN SHPVQVGRII LEDYHDHGLL RVRMVNLQVE DSGLYQCVIY QPPKEPHMLF DRIRLVVTKG FSGTPGSNEN STQNVYKIPP TTTKALCPLY TSPRTVTQAP PKSTADVSTP DSEINLTNVT DIIRVPVFN
Videos
SDS
Documents
Certificates