missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TREM2 Protein

A DNA sequence encoding the Homo sapiens (Human) TREM2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6839-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046200

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001258750.1
TREM2 Protein
Research Use Only
Trem2
Recombinant Protein
E. coli
HNTTVFQGVA GQSLQVSCPY DSMKHWGRRK AWCRQLGEKG PCQRVVSTHN LWLLSFLRRW NGSTAITDDT LGGTLTITLR NLQPHDAGLY QCQSLHGSEA DTLRKVLVEV LADPLDHRDA GDLWFPGESE SFEDAHVEHS ISRSLLEGEI PFPPTS
Greater than 90% as determined by SDS-PAGE.
54209
10 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers