missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trimethyllysine dioxygenase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Trimethyllysine dioxygenase, mitochondrial, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001171726.1 |
---|---|
Gene ID (Entrez) | 55217 |
Name | Trimethyllysine dioxygenase, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035990
|
enQuireBio™
QP6808-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055990
|
enQuireBio™
QP6808-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045990
|
enQuireBio™
QP6808-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065990
|
enQuireBio™
QP6808-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075990
|
enQuireBio™
QP6808-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085990
|
enQuireBio™
QP6808-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001171726.1 | |
Trimethyllysine dioxygenase, mitochondrial Protein | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55217 | |
Research Use Only | |
TMLHE | |
Recombinant Protein | |
E. coli | |
LLKGGVIYPA LPQPNFKSLL PLAVHWHHTA SKSLTCAWQQ HEDHFELKYA NTVMRFDYVW LRDHCRSASC YNSKTHQRSL DTASVDLCIK PKTIRLDETT LFFTWPDGHV TKYDLNWLVK NSYEGQKQKV IQPRILWNAE IYQQAQVPSV DCQSFLETNE GLKKFLQNFL LYGIAFVENV PPTQEHTEKL AERISLIRET IYGRMWYFTS DFSRGDTAYT KLALDRHTDT TYFQEPCGIQ VFHCLKHEGT GGRTLLVDGF YAAEQVLQKA PEEFELLSKV PLKHEYIEDV GECHNHMIGI GPVLNIYPWN KELYLIRLFK EKQNTVNRQW NSSLQCDIPE RILTYRHFVS GTSIEHRGSL I | |
Greater than 90% as determined by SDS-PAGE. |