missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Troponin IC Protein
A cDNA sequence encoding the Troponin IC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13818-1mg
Specifications
Troponin IC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Each mg/ml of protein (0.5 mg/ml) contains 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |