missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trypsin-2 Protein
A cDNA sequence encoding the Trypsin-2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 725.00 €
Specifications
Name | Trypsin-2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945508
|
enQuireBio™
QP10899-1MG |
1 mg |
131.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965508
|
enQuireBio™
QP10899-5MG |
5 mg |
203.00 €
5mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955508
|
enQuireBio™
QP10899-50MG |
50 mg |
725.00 €
50mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Trypsin-2 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
The protein was lyophilized without any additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MNLLLILTFV AAAVAAPFDD DDKIVGGYIC EENSVPYQVS LNSGYHFCGG SLISEQWVVS AGHCYKSRIQ VRLGEHNIEV LEGNEQFINA AKIIRHPKYN SRTLDNDILL IKLSSPAVIN SRVSAISLPT APPAAGTESL ISGWGNTLSS GADYPDELQC LDAPVLSQAE CEASYPGKIT NNMFCVGFLE GGKDSCQGDS GGPVVSNGEL QGIVSWGYGC AQKNRPGVYT KVYNYVDWIK DTIAANS | |
Greater than 90% as determined by SDS-PAGE. |