missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TTC32 Protein
A cDNA sequence encoding the TTC32 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | TTC32 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994859
|
enQuireBio™
QP13837-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984859
|
enQuireBio™
QP13837-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974859
|
enQuireBio™
QP13837-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TTC32 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TTC32 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 2mM DTT and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEGQRQE SHATLTLAQA HFNNGEYAEA EALYSAYIRR CACAASSDES PGSKCSPEDL ATAYNNRGQI KYFRVDFYEA MDDYTSAIEV QPNFEVPYYN RGLILYRLGY FDDALEDFKK VLDLNPGFQD ATLSLKQTIL DKEEKQRRNV AKNY | |
Greater than 90% as determined by SDS-PAGE. |