missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TTC32 Protein

A cDNA sequence encoding the TTC32 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13837-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984859

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

TTC32 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TTC32 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 2mM DTT and 20% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEGQRQE SHATLTLAQA HFNNGEYAEA EALYSAYIRR CACAASSDES PGSKCSPEDL ATAYNNRGQI KYFRVDFYEA MDDYTSAIEV QPNFEVPYYN RGLILYRLGY FDDALEDFKK VLDLNPGFQD ATLSLKQTIL DKEEKQRRNV AKNY
Videos
SDS
Documents
Certificates