missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TXNRD1 / TRXR1 Protein
A DNA sequence encoding the Homo sapiens (Human) TXNRD1 / TRXR1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001087240.1 |
---|---|
Gene ID (Entrez) | 7296 |
Name | TXNRD1 / TRXR1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029260
|
enQuireBio™
QP7539-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049260
|
enQuireBio™
QP7539-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039260
|
enQuireBio™
QP7539-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059260
|
enQuireBio™
QP7539-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069260
|
enQuireBio™
QP7539-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079260
|
enQuireBio™
QP7539-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001087240.1 | |
TXNRD1 / TRXR1 Protein | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 5 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7296 | |
Research Use Only | |
TXNRD1 | |
Recombinant Protein | |
E. coli | |
MNGPEDLPKS YDYDLIIIGG GSGGLAAAKE AAQYGKKVMV LDFVTPTPLG TRWGLGGTCV NVGCIPKKLM HQAALLGQAL QDSRNYGWKV EETVKHDWDR MIEAVQNHIG SLNWGYRVAL REKKVVYENA YGQFIGPHRI KATNNKGKEK IYSAERFLIA TGERPRYLGI PGDKEYCISS DDLFSLPYCP GKTLVVGASY VALECAGFLA GIGLDVTVMV RSILLRGFDQ DMANKIGEHM EEHGIKFIRQ FVPIKVEQIE AGTPGRLRVV AQSTNSEEII EGEYNTVMLA IGRDACTRKI GLETVGVKIN EKTGKIPVTD EEQTNVPYIY AIGDILEDKV ELTPVAIQAG RLLAQRLYAG STVKCDYENV PTTVFTPLEY GACGLSEEKA VEKFGEENIE VYHSYFWPLE WTIPSRDNNK CYAKIICNTK DNERVVGFHV LGPNAGEVTQ GFAAALKCGL TKKQLDSTIG IHPVCAEVFT TLSVTKRSGA SILQAGC | |
Greater than 90% as determined by SDS-PAGE. |