missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TXNRD3NB Protein
A cDNA sequence encoding the TXNRD3NB was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Gene ID (Entrez) | 645840 |
---|---|
Name | TXNRD3NB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | TXNRD3NB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974879
|
enQuireBio™
QP13843-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964879
|
enQuireBio™
QP13843-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954879
|
enQuireBio™
QP13843-0.1MG |
100 μg |
1740.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
645840 | |
Research Use Only | |
TXNRD3NB | |
Human | |
His | |
TXNRD3NB protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0), 30% glycerol, 0.15M NaCl and 1mM DTT. |
TXNRD3NB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMRDLSER RLGQPELKAE QQMPLEPVRA RLSVGLACCC SHTTAEASSL EHGDKVFGQG FPSPLEEIKR LLKISRALQA RSVPSTQEKA KCLSGEPGQP EGKGQETYPG PGKVEGKAEP AMRKDDVCPG MKCISG | |
Greater than 85.0% as determined by SDS-PAGE. |