missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tyrosinase Protein
A DNA sequence encoding the Homo sapiens (Human) Tyrosinase, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_000363.1 |
---|---|
Gene ID (Entrez) | 7299 |
Name | Tyrosinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026360
|
enQuireBio™
QP6860-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066360
|
enQuireBio™
QP6860-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036360
|
enQuireBio™
QP6860-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056360
|
enQuireBio™
QP6860-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076360
|
enQuireBio™
QP6860-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046360
|
enQuireBio™
QP6860-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000363.1 | |
Tyrosinase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7299 | |
Research Use Only | |
TYR | |
Recombinant Protein | |
Yeast | |
HFPRACVSSK NLMEKECCPP WSGDRSPCGQ LSGRGSCQNI LLSNAPLGPQ FPFTGVDDRE SWPSVFYNRT CQCSGNFMGF NCGNCKFGFW GPNCTERRLL VRRNIFDLSA PEKDKFFAYL TLAKHTISSD YVIPIGTYGQ MKNGSTPMFN DINIYDLFVW MHYYVSMDAL LGGSEIWRDI DFAHEAPAFL PWHRLFLLRW EQEIQKLTGD ENFTIPYWDW RDAEKCDICT DEYMGGQHPT NPNLLSPASF FSSWQIVCSR LEEYNSHQSL CNGTPEGPLR RNPGNHDKSR TPRLPSSADV EFCLSLTQYE SGSMDKAANF SFRNTLEGFA SPLTGIADAS QSSMHNALHI YMNGTMSQV | |
Greater than 90% as determined by SDS-PAGE. |