missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TYW5 Protein
A cDNA sequence encoding the TYW5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13847-20ug
Additional Details : Weight : 0.01000kg
Specifications
TYW5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGQHLP VPRLEGVSRE QFMQHLYPQR KPLVLEGIDL GPCTSKWTVD YLSQVGGKKE VKIHVAAVAQ MDFISKNFVY RTLPFDQLVQ RAAEEKHKEF FVSEDEKYYL RSLGEDPRKD VADIRKQFPL LKGDIKFPEF FKEEQFFSSV FRISSPGLQL WTHYDVMDNL LIQVTGKKRV VLFSPRDAQY LYLKGTKSEV LNIDNPDLAK YPLFSKARRY ECSLEAGDVL FIPALWFHNV ISEEFGVGVN IFWKHLPSEC YDKTDTYGNK DPTAASRAAQ ILDRALKTLA ELPEEYRDFY ARRMVLHIQD KAYSKNSE | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TYW5 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |