missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBE2B Protein

A cDNA sequence encoding the UBE2B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13853-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984909

  • 6549.36 € / 1mg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UBE2B Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Lyophilized from a 0.2?m filtered concentrated (1 mg/ml) solution in 1X PBS and 1mM DTT, pH 7.5.
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MHHHHHHAMGQLRSMSTPARRRLMRDFKRLQEDPPVGVSGAPSENNIMQWNAVIFGPEGTPFEDGTFKLVIEFSEEYPNKPPTVRFLSKMFHPNVYADGSICLDILQNRWSPTYDVSSILTSIQSLLDEPNPNSPANSQAAQLYQENKREYEKRVSAIVEQSWNDS
Videos
SDS
Documents
Certificates