missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBE2L3 Protein

A cDNA sequence encoding the UBE2L3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13863-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984939

Quantity Price
1 2269.63 € / 1mg
3 2133.68 € / 1mg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

UBE2L3 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein contains 50mM HEPES (pH 7.5) 150mM NaCl, 1mM DTT, and 10% glycerol.
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MAASRRLMKE LEEIRKCGMK NFRNIQVDEA NLLTWQGLIV PDNPPYDKGA FRIEINFPAE YPFKPPKITF KTKIYHPNID EKGQVCLPVI SAENWKPATK TDQVIQSLIA LVNDPQPEHP LRADLAEEYS KDRKKFCKNA EEFTKKYGEK RPVD
Videos
SDS
Documents
Certificates