missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBLCP1 Protein

A cDNA sequence encoding the UBLCP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13875-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984979

  • 6549.36 € / 1mg

Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UBLCP1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
UBLCP1 protein solution (0.25 mg/ml) in Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMALPIIV KWGGQEYSVT TLSEDDTVLD LKQFLKTLTG VLPERQKLLG LKVKGKPAEN DVKLGALKLK PNTKIMMMGT REESLEDVLG PPPDNDDVVN DFDIEDEVVE VENREENLLK ISRRVKEYKV EILNPPREGK KLLVLDVDYT LFDHRSCAET GVELMRPYLH EFLTSAYEDY DIVIWSATNM KWIEAKMKEL GVSTNANYKI TFMLDSAAMI TVHTPRRGLI DVKPLGVIWG KFSEFYSKKN TIMFDDIGRN FLMNPQNGLK IRPFMKAHLN RDKDKELLKL TQYLKEIAKL DDFLDLNHKY WERYLSKKQG Q
Videos
SDS
Documents
Certificates