missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBXN2B Protein
A cDNA sequence encoding the UBXN2B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13878-2ug
Additional Details : Weight : 0.01000kg
Specifications
UBXN2B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEGGGP EPGEQERRSS GPRPPSARDL QLALAELYED EVKCKSSKSN RPKATVFKSP RTPPQRFYSS EHEYSGLNIV RPSTGKIVNE LFKEAREHGA VPLNEATRAS GDDKSKSFTG GGYRLGSSFC KRSEYIYGEN QLQDVQILLK LWSNGFSLDD GELRPYNEPT NAQFLESVKR GEIPLELQRL VHGGQVNLDM EDHQDQEYIK PRLRFKAFSG EGQKLGSLTP EIVSTPSSPE EEDKSILNAV VLIDDSVPTT KIQIRLADGS RLIQRFNSTH RILDVRNFIV QSRPEFAALD FILVTSFPNK ELTDESLTLL EADILNTVLL QQLK | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UBXN2B solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |