missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ULBP1 Protein

A cDNA sequence encoding the ULBP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13885-HIS-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985009

  • 131.00 € / 2µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ULBP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMGWVDT HCLCYDFIIT PKSRPEPQWC EVQGLVDERP FLHYDCVNHK AKAFASLGKK VNVTKTWEEQ TETLRDVVDF LKGQLLDIQV ENLIPIEPLT LQARMSCEHE AHGHGRGSWQ FLFNGQKFLL FDSNNRKWTA LHPGAKKMTE KWEKNRDVTM FFQKISLGDC KMWLEEFLMY WEQMLDPTKP PSLAPG
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ULBP1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 150mM NaCl and 30% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers