missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Uncharacterized protein C4orf3 Protein

A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C4orf3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7860-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051231

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001001701.2
Uncharacterized protein C4orf3 Protein
Research Use Only
C4orf3
Recombinant Protein
E. coli
MEVDAPGVDG RDGLRERRGF SEGGRQNFDV RPQSGANGLP KHSY
Greater than 90% as determined by SDS-PAGE.
401152
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers