missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Urokinase Protein

A cDNA sequence encoding the Urokinase was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13908-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955089

  • 179.00 € / 1mg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

Urokinase Protein
Research Use Only
Recombinant Protein
Human
The Urokinase was lyophilized from a concentrated (1 mg/ml) solution containing phosphate buffer.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Greater than 94.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers