missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human USP15 Protein
A cDNA sequence encoding the USP15 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | USP15 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955099
|
enQuireBio™
QP13911-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935099
|
enQuireBio™
QP13911-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945099
|
enQuireBio™
QP13911-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
USP15 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The USP15 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEGGAA DLDTQRSDIA TLLKTSLRKG DTWYLVDSRW FKQWKKYVGF DSWDKYQMGD QNVYPGPIDN SGLLKDGDAQ SLKEHLIDEL DYILLPTEGW NKLVSWYTLM EGQEPIARKV VEQGMFVKHC KVEVYLTELK LCENGNMNNV VTRRFSKADT IDTIEKEIRK IFSIPDEKET RLWNKYMSNT FEPLNKPDST IQDAGLYQGQ VLVIEQKNED GTWPRGPSTP KKPLEQSC | |
Greater than 90% as determined by SDS-PAGE. |