missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UTI Protein
A cDNA sequence encoding the UTI was constructed and used to recombinantly synthesize the protein.
131.00 € - 216.00 €
Specifications
Name | UTI Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Human UTI has an activity of 2390IU/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955538
|
enQuireBio™
QP10909-2.5MG |
2.5 mg |
131.00 €
2.50mg |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965538
|
enQuireBio™
QP10909-5MG |
5 mg |
131.00 €
5mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945538
|
enQuireBio™
QP10909-10MG |
10 mg |
216.00 €
10mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
UTI Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human | |
Lyophilized from a (1 mg/ml) solution containing no additives. |
Research Use Only | |
Human UTI has an activity of 2390IU/mg. | |
Human | |
Untagged |