missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vasculin Protein
A DNA sequence encoding the Homo sapiens (Human) Vasculin, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001120707.2 |
---|---|
Gene ID (Entrez) | 65056 |
Name | Vasculin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050841
|
enQuireBio™
QP7795-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16070841
|
enQuireBio™
QP7795-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060841
|
enQuireBio™
QP7795-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080841
|
enQuireBio™
QP7795-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090841
|
enQuireBio™
QP7795-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000851
|
enQuireBio™
QP7795-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001120707.2 | |
Vasculin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
65056 | |
Research Use Only | |
GPBP1 | |
Recombinant Protein | |
E. coli | |
MRTDKKSEFL KALKRDRVEE EHEDESRAGS EKDDDSFNLH NSNSTHQERD INRNFDENEI PQENGNASVI SQQIIRSSTF PQTDVLSSSL EAEHRLLKEM GWQEDSENDE TCAPLTEDEM REFQVISEQL QKNGLRKNGI LKNGLICDFK FGPWKNSTFK PTTENDDTET SSSDTSDDDD V | |
Greater than 90% as determined by SDS-PAGE. |