missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VDR Protein

A cDNA sequence encoding the VDR was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13932-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985159

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

VDR Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Flag Tag
VDR protein is supplied in 50mM Tris, 150mM NaCl and 10% Glycerol, pH 7.5.
Greater than 70.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MSYYHHHHHHDYDIPTTDYKDDDDKDYKDDDDKENLYFQGEFMEAMAASTSLPDPGDFDRNVPRICGVCGDRATGFHFNAMTCEGCKGFFRRSMKRKALFTCPFNGDCRITKDNRRHCQACRLKRCVDIGMMKEFILTDEEVQRKREMILKRKEEEALKDSLRPKLSEEQQRIIAILLDAHHKTYDPTYSDFCQFRPPVRVNDGGGSHPSRPNSRHTPSFSGDSSSSCSDHCITSSDMMDSSSFSNLDLSEEDSDDPSVTLELSQLSMLPHLADLVSYSIQKVIGFAKMIPGFRDLTSEDQIVLLKSSAIEVIMLRSNESFTMDDMSWTCGNQDYKYRVSDVTKAGHSLELIEPLIKFQVGLKKLNLHEEEHVLLMAICIVSPDRPGVQDAALIEAIQDRLSNTLQTYIRCRHPPPGSHLLYAKMIQKLADLRSLNEEHSKQYRCLSFQPECSMKLTPLVLEVFGNEIS
Videos
SDS
Documents
Certificates