missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VOPP1 Protein
A cDNA sequence encoding the VOPP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13939-1mg
Specifications
VOPP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRRRMYPPPL IEEPAFNVSY TRQPPNPGPG AQQPGPPYYT DPGGPGMNPV GNSMAMAFQV PPNSPQGSVA CPPPPAYCNT PPPPYEQVVK AK | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VOPP1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.2M NaCl. |