missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VPS26A Protein

A cDNA sequence encoding the VPS26A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13942-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985189

Quantity Price
1 114.77 € / 2µg
10 107.33 € / 2µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

9559
Greater than 85% as determined by SDS-PAGE.
Research Use Only
VPS26A
Human
His
VPS26A protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.15M NaCl.
VPS26A Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSFLGGF FGPICEIDIV LNDGETRKMA EMKTEDGKVE KHYLFYDGES VSGKVNLAFK QPGKRLEHQG IRIEFVGQIE LFNDKSNTHE FVNLVKELAL PGELTQSRSY DFEFMQVEKP YESYIGANVR LRYFLKVTIV RRLTDLVKEY DLIVHQLATY PDVNNSIKME VGIEDCLHIE FEYNKSKYHL KDVIVGKIYF LLVRIKIQHM ELQLIKKEIT GIGPSTTTET ETIAKYEIMD GAPVKGESIP IRLFLAGYDP TPTMRDVNKK FSVRYFLNLV LVDEEDRRYF KQQEIILWRK APEKLRKQRT NFHQRFESPE SQASAEQPEM
Videos
SDS
Documents
Certificates