missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VPS4B Protein
A cDNA sequence encoding the VPS4B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13946-10ug
Additional Details : Weight : 0.01000kg
Specifications
VPS4B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSSTSP NLQKAIDLAS KAAQEDKAGN YEEALQLYQH AVQYFLHVVK YEAQGDKAKQ SIRAKCTEYL DRAEKLKEYL KNKEKKAQKP VKEGQPSPAD EKGNDSDGEG ESDDPEKKKL QNQLQGAIVI ERPNVKWSDV AGLEGAKEAL KEAVILPIKF PHLFTGKRTP WRGILLFGPP GTGKSYLAKA VATEANNSTF FSISSSDLVS KWLGESEKLV KNLFQLAREN KPSIIFIDEI DSLCGSRSEN ESEAARRIKT EFLVQMQGVG VDNDGILVLG ATNIPWVLDS AIRRRFEKRI YIPLPEPHAR AAMFKLHLGT TQNSLTEADF RELGRKTDGY SGADISIIVR DALMQPVRKV QSATHFKKVR GPSRADPNHL VDDLLTPCSP GDPGAIEMTW MDVPGDKLLE PVVSMSDMLR SLSNTKPTVN EHDLLKLKKF TEDFGQEG | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
VPS4B protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol, 1mM DTT. |