missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WFDC2 Protein
A cDNA sequence encoding the WFDC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13956-10ug
Additional Details : Weight : 0.01000kg
Specifications
WFDC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASEKTGVCPELQ ADQNCTQECV SDSECADNLK CCSAGCATFC SLPNDKEGSC PQVNINFPQL GLCRDQCQVD SQCPGQMKCC RNGCGKVSCV TPNF | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
WFDC2 filtered (0.4microm) and lyophilized from 0.5 mg/ml in PBS buffer, pH 7.5. |