missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human X-ray repair cross-complementing protein 1
A DNA sequence encoding the Homo sapiens (Human) X-ray repair cross-complementing protein 6, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001275905.1 |
---|---|
Gene ID (Entrez) | 2547 |
Name | X-ray repair cross-complementing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044301
|
enQuireBio™
QP8378-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064301
|
enQuireBio™
QP8378-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054301
|
enQuireBio™
QP8378-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074301
|
enQuireBio™
QP8378-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084301
|
enQuireBio™
QP8378-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094301
|
enQuireBio™
QP8378-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001275905.1 | |
X-ray repair cross-complementing protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2547 | |
Research Use Only | |
XRCC6 | |
Recombinant Protein | |
E. coli | |
SYYKTEGDEE AEEEQEENLE ASGDYKYSGR DSLIFLVDAS KAMFESQSED ELTPFDMSIQ CIQSVYISKI ISSDRDLLAV VFYGTEKDKN SVNFKNIYVL QELDNPGAKR ILELDQFKGQ QGQKRFQDMM GHGSDYSLSE VLWVCANLFS DVQFKMSHKR IMLFTNEDNP HGNDSAKASR ARTKAGDLRD TGIFLDLMHL KKPGGFDISL FYRDIIS | |
Greater than 80% as determined by SDS-PAGE. |