missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human XG Protein
A cDNA sequence encoding the XG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13971-2ug
Additional Details : Weight : 0.01000kg
Specifications
7499 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
XG | |
Human | |
His | |
XG protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
XG Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQRDFDLA DALDDPEPTK KPNSDIYPKP KPPYYPQPEN PDSGGNIYPR PKPRPQPQPG NSGNSGGYFN DVDRDDGRYP PRPRPRPPAG GGGGGYSSYG NSDNTHGGDH HSTYGNPEGN MVAK |