missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein GLI2 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein GLI2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6088-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989869

Quantity Price
1 434.28 € / 100µg
6 408.22 € / 100µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_005261.2
Zinc finger protein GLI2 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
2736
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GLI2
Recombinant Protein
E. coli
EQLADLKEDL DRDDCKQEAE VVIYETNCHW EDCTKEYDTQ EQLVHHINNE HIHGEKKEFV CRWQACTREQ KPFKAQYMLV VHMRRHTGEK PHKCTFEGCS KAYSRLENLK THLRSHTGEK PYVCEHEGCN KAFSNASDRA KHQNRTHSNE KPYICKIPGC TKRYTDPSSL RKHVKTVHGP DAHVTKKQRN DVHLRTPLLK ENGDSEAGTE PGGPESTEAS STSQAVEDCL
Videos
SDS
Documents
Certificates