missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZRANB2 Protein
A DNA sequence encoding the Homo sapiens (Human) ZRANB2, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_005446.2 |
---|---|
Gene ID (Entrez) | 9406 |
Name | ZRANB2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036710
|
enQuireBio™
QP6912-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056710
|
enQuireBio™
QP6912-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046710
|
enQuireBio™
QP6912-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066710
|
enQuireBio™
QP6912-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076710
|
enQuireBio™
QP6912-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086710
|
enQuireBio™
QP6912-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005446.2 | |
ZRANB2 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9406 | |
Research Use Only | |
ZRANB2 | |
Recombinant Protein | |
E. coli | |
MSTKNFRVSD GDWICPDKKC GNVNFARRTS CNRCGREKTT EAKMMKAGGT EIGKTLAEKS RGLFSANDWQ CKTCSNVNWA RRSECNMCNT PKYAKLEERT GYGGGFNERE NVEYIEREES DGEYDEFGRK KKKYRGKAVG PASILKEVED KESEGEEEDE DEDLSKYKLD EDEDEDDADL SKYNLDASEE EDSNKKKSNR RSRSKSRSSH SRSSSRSSSP SSSRSRSRSR SRSSSSSQSR SRSSSRERSR SRGSKSRSSS RSHRGSSSPR KRSYSSSSSS PERNRKRSRS RSSSSGDRKK RRTRSRSPES QVIGENTKQP | |
Greater than 90% as determined by SDS-PAGE. |