missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zymogen granule protein 16 homolog B Protein
A DNA sequence encoding the Homo sapiens (Human) Zymogen granule protein 16 homolog B, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_660295.2 |
---|---|
Gene ID (Entrez) | 124220 |
Name | Zymogen granule protein 16 homolog B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16001281
|
enQuireBio™
QP7869-YE-100UG |
100 μg |
880.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041281
|
enQuireBio™
QP7869-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011281
|
enQuireBio™
QP7869-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031281
|
enQuireBio™
QP7869-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051281
|
enQuireBio™
QP7869-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021281
|
enQuireBio™
QP7869-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_660295.2 | |
Zymogen granule protein 16 homolog B Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
124220 | |
Research Use Only | |
ZG16B | |
Recombinant Protein | |
Yeast | |
GKMYGPGGGK YFSTTEDYDH EITGLRVSVG LLLVKSVQVK LGDSWDVKLG ALGGNTQEVT LQPGEYITKV FVAFQAFLRG MVMYTSKDRY FYFGKLDGQI SSAYPSQEGQ VLVGIYGQYQ LLGIKSIGFE WNYPLEEPTT EPPVNLTYSA NSPVGR | |
Greater than 90% as determined by SDS-PAGE. |