missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Hong Kong Recombinant Protein
A cDNA sequence encoding the H3N2 Hong Kong Recombinant was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | Influenza H3N2 Hong Kong Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15999308
|
enQuireBio™
QP12082-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989308
|
enQuireBio™
QP12082-10UG |
10 μg |
288.00 €
10µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979308
|
enQuireBio™
QP12082-100UG |
100 μg |
2610.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H3N2 Hong Kong Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H3N2 A/ Hong Kong 4801/2014 solution contains 10mM Sodium phosphate, pH 7.4,150mM NaCl and 0.005% Tween-20. |