missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Kiev Protein
Recombinant Protein
131.00 € - 2900.00 €
Specifications
Name | Influenza H3N2 Kiev Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909318
|
enQuireBio™
QP12083-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929318
|
enQuireBio™
QP12083-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919318
|
enQuireBio™
QP12083-1MG |
1 mg |
2900.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H3N2 Kiev Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The H3N2 A/Kiev/301/94 solution contains 0.1M NaCl, 10mM Tris-Hcl, 1mM EDTA pH 8.0, 0.1% sodium azide (NaN3) and 0.005% thimerosal. |