missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H7N7 Netherlands Protein

A cDNA sequence encoding the H7N7 Netherlands was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12095-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989348

Quantity Price
1 2269.63 € / 100µg
3 2133.68 € / 100µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

Influenza H7N7 Netherlands Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
The Recombinant H7N7 A/Netherlands/219/03 solution contains 10mM Sodium phosphate, pH 7 and 150mM NaCl.
Videos
SDS
Documents
Certificates