missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza New York 3571/2009 Protein

A cDNA sequence encoding the New York 3571/2009 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12833-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991689

Quantity Price
1 114.77 € / 2µg
10 107.33 € / 2µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

Influenza New York 3571/2009 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
His
The H1N1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Baculovirus
ADLMDTLCIG YHANNSTDTV DTVLEKNVTV THSVNLLEDK HNGKLCKLRG VAPLHLGKCN IAGWILGNPE CESLSTASSW SYIVETSSSD NGTCYPGDFI DYEELREQLS SVSSFERFEI FPKTSSWPNH DSNKGVTAAC PHAGAKSFYK NLIWLVKKGN SYPKLSKSYI NDKGKEVLVL WGIHHPSTSA DQQSLYQNAD AYVFVGSSRY SKKFKPEIAI RPKVRDQEGR MNYYWTLVEP GDKITFEATG NLVVPRYAFA MERNAGSGII ISDTPVHDCN TTCQTPKGAI NTSLPFQNIH PITIGKCPKY VKSTKLRLAT GLRNVPSIQS RSRHHHHHH
Videos
SDS
Documents
Certificates