missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Signal peptidase I Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Signal peptidase I, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7516-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039130

  • 967.00 € / 100µg
Estimated Shipment: 05-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_217419.1
M. tuberculosis Signal peptidase I Protein
100 μg
Not determined.
Extracellular Domain
M. tuberculosis
His-SUMO
Tris-based buffer, 50% glycerol
887157
Greater than 90% as determined by SDS-PAGE.
Research Use Only
lepB
Recombinant Protein
E. coli
RPYLIPSESM EPTLHGCSTC VGDRIMVDKL SYRFGSPQPG DVIVFRGPPS WNVGYKSIRS HNVAVRWVQN ALSFIGFVPP DENDLVKRVI AVGGQTVQCR SDTGLTVNGR PLKEPYLDPA TMMADPSIYP CLGSEFGPVT VPPGRVWVMG DNRTHSADSR AHCPLLCTDD PLPGTVPVAN VIGKARLIVW PPSRWGVVRS VNPQQGR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers