missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL-15-S48AA Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-15-S48AA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9208-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001038196.1 | |
IL-15-S48AA Protein | |
Research Use Only | |
IL15 | |
Recombinant Protein | |
Yeast | |
NWVNVISDLK KIEDLIQSMH IDATLYTESD VHPSCKVTAM KCFLLELQVI SHESGDTDIH DTVENLIILA NNILSSNGNI TESGCKECEE LEEKNIKEFL QSFVHIVQMF INTS | |
Greater than 90% as determined by SDS-PAGE. |
699616 | |
50 μg | |
Not determined. | |
Full Length | |
Macaque | |
His | |
Tris-based buffer, 50% glycerol |