missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse 60S acidic ribosomal protein P0 Protein
A DNA sequence encoding the Mus musculus (Mouse) 60S acidic ribosomal protein P0, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_031501.1 |
---|---|
Gene ID (Entrez) | 11837 |
Name | 60S acidic ribosomal protein P0 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044630
|
enQuireBio™
QP6624-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064630
|
enQuireBio™
QP6624-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054630
|
enQuireBio™
QP6624-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074630
|
enQuireBio™
QP6624-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084630
|
enQuireBio™
QP6624-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094630
|
enQuireBio™
QP6624-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_031501.1 | |
60S acidic ribosomal protein P0 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11837 | |
Research Use Only | |
RPLP0 | |
Recombinant Protein | |
E. coli | |
MPREDRATWK SNYFLKIIQL LDDYPKCFIV GADNVGSKQM QQIRMSLRGK AVVLMGKNTM MRKAIRGHLE NNPALEKLLP HIRGNVGFVF TKEDLTEIRD MLLANKVPAA ARAGAIAPCE VTVPAQNTGL GPEKTSFFQA LGITTKISRG TIEILSDVQL IKTGDKVGAS EATLLNMLNI SPFSFGLIIQ QVFDNGSIYN PEVLDITEQA LHSRFLEGVR NVASVCLQIG YPTVASVPHS IINGYKRVLA LSVETEYTFP LTEKVKAFLA DPSAFAAAAP AAAATTAAPA AAAAPAKAEA KEESEESDED MGFGLFD | |
Greater than 80% as determined by SDS-PAGE. |