missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Acetylcholine receptor subunit alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) Acetylcholine receptor subunit alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5833-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031415.2 | |
Acetylcholine receptor subunit alpha Protein | |
Research Use Only | |
Chrna1 | |
Recombinant Protein | |
E. coli | |
SEHETRLVAK LFEDYSSVVR PVEDHREIVQ VTVGLQLIQL INVDEVNQIV TTNVRLKQQW VDYNLKWNPD DYGGVKKIHI PSEKIWRPDV VLYNNADGDF AIVKFTKVLL DYTGHITWTP PAIFKSYCEI IVTHFPFDEQ NCSMKLGTWT YDGSVVAINP ESDQPDLSNF MESGEWVIKE ARGWKHWVFY SCCPTTPYLD ITYHFVMQRL | |
Greater than 80% as determined by SDS-PAGE. |
11435 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |