missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AHSG Protein
A cDNA sequence encoding the AHSG was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | AHSG Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955778
|
enQuireBio™
QP10984-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935778
|
enQuireBio™
QP10984-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945778
|
enQuireBio™
QP10984-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
AHSG Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
AHSG protein solution (0.25 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
APQGTGLGFR ELACDDPEAE QVALLAVDYL NNHLLQGFKQ VLNQIDKVKV WSRRPFGVVY EMEVDTLETT CHALDPTPLA NCSVRQLTEH AVEGDCDFHI LKQDGQFRVM HTQCHSTPDS AEDVRKLCPR CPLLTPFNDT NVVHTVNTAL AAFNTQNNGT YFKLVEISRA QNVPLPVSTL VEFVIAATDC TAKEVTDPAK CNLLAEKQHG FCKANLMHNL GGEEVSVACK LFQTQPQPAN ANAVGPVPTA NAALPADPPA SVVVGPVVVP RGLSDHRTYH DLRHAFSPVA SVESASGETL HSPKVGQPGA AGPVSPMCPG RIRHFKIHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |