missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Alkaline Phosphatase / ALPL Protein
A DNA sequence encoding the Mus musculus (Mouse) Alkaline Phosphatase / ALPL, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001274101.1 |
---|---|
Gene ID (Entrez) | 11647 |
Name | Alkaline Phosphatase / ALPL Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016981
|
enQuireBio™
QP8885-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036981
|
enQuireBio™
QP8885-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026981
|
enQuireBio™
QP8885-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046981
|
enQuireBio™
QP8885-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056981
|
enQuireBio™
QP8885-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066981
|
enQuireBio™
QP8885-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001274101.1 | |
Alkaline Phosphatase / ALPL Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
11647 | |
Research Use Only | |
Alpl | |
Recombinant Protein | |
E. coli | |
PEKERDPSYW RQQAQETLKN ALKLQKLNTN VAKNVIMFLG DGMGVSTVTA ARILKGQLHH NTGEETRLEM DKFPFVALSK TYNTNAQVPD SAGTATAYLC GVKANEGTVG VSAATERTRC NTTQGNEVTS ILRWAKDAGK SVGIVTTTRV NHATPSAAYA HSADRDWYSD NEMPPEALSQ GCKDIAYQLM HNIKDIDVIM GGGRKYMYPK NRTDVEYELD EKARGTRLDG LDLISIWKSF KPRHKHSHYV WNRTELLALD PSRVDYLLGL FEPGDMQYEL NRNNLTDPSL SEMVEVALRI LTKNLKGFFL LVEGGRIDHG HHEGKAKQAL HEAVEMDQAI GKAGAMTSQK DTLTVVTADH SHVFTFGGYT PRGNSIFGLA PMVSDTDKKP FTAILYGNGP GYKVVDGERE NVSMVDYAHN NYQAQSAVPL RHETHGGEDV AVFAKGPMAH LLHGVHEQNY IPHVMAYASC IGANLDHCAW AGSG | |
Greater than 90% as determined by SDS-PAGE. |