missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Alpha-defensin 5 Protein
A DNA sequence encoding the Mus musculus (Mouse) Alpha-defensin 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5925-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Alpha-defensin 5 Protein | |
Research Use Only | |
Defa5 | |
Recombinant Protein | |
E. coli | |
SKKLICYCRI RGCKRRERVF GTCRNLFLTF VFCCS | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |