missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ANTXR2 Protein
A cDNA sequence encoding the ANTXR2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11030-50ug
Additional Details : Weight : 0.01000kg
Specifications
ANTXR2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
QAQEQPSCKK AFDLYFVLDK SGSVANNWIE IYNFVHQLTE RFVSPEMRLS FIVFSSQATI ILPLTGDRYK IGKGLEDLKA VKPVGETYIH EGLKLANEQI QNAGGLKASS IIIALTDGKL DGLVPSYAEN EAKKSRSLGA SVYCVGVLDF EQAQLERIAD SKDQVFPVKG GFQALKGIIN SILAQSCTEI LELSPSSVCV GEKFQVVLTG RAVTSISHDG SVLCTFTANS TYTKSEKPVS IQPSSILCPA PVLNKDGETL EVSISYNDGK SAVSRSLTIT ATECTNGLEH HHHHH | |
Greater than 95% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
ANTXR2 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |