missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Apolipoprotein 1
A DNA sequence encoding the Mus musculus (Mouse) Apolipoprotein E, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5668-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001292748.1 | |
Apolipoprotein 1 | |
Research Use Only | |
Apoe | |
Recombinant Protein | |
E. coli | |
EGEPEVTDQL EWQSNQPWEQ ALNRFWDYLR WVQTLSDQVQ EELQSSQVTQ ELTALMEDTM TEVKAYKKEL EEQLGPVAEE TRARLGKEVQ AAQARLGADM EDLRNRLGQY RNEVHTMLGQ STEEIRARLS THLRKMRKRL MRDAEDLQKR LAVYKAGARE GAERGVSAIR ERLGPLVEQG RQRTANLGAG AAQPLRDRAQ AFGDRIRGRL EEVGNQARDR LEEVREHMEE VRSKMEEQTQ QIRLQAEIFQ ARLKGWFEPI VEDMHRQWAN LMEKIQASVA TNPIITPVAQ ENQ | |
Greater than 89.4% as determined by SDS-PAGE. |
11816 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |