missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse BCA 1 Protein

A cDNA sequence encoding the BCA 1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10530-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984148

  • 3401.86 € / 1mg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

55985
Greater than 97.0% as determined by SDS-PAGE.
Research Use Only
Cxcl13
Recombinant Protein
E. coli
ILEAHYTNLK CRCSGVISTV VGLNIIDRIQ VTPPGNGCPK TEVVIWTKMK KVICVNPRAK WLQRLLRHVQ SKSLSSTPQA PVSKRRAA
BCA 1 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50, as measured by its ability to chemoattract human CXCR5-transfected mouse BaF3 cells, is less than 2microg/ml.
Mouse
Untagged
The BCA1 protein was lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA.
Videos
SDS
Documents
Certificates