missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Bcl-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Bcl-2, was expressed in the hosts and tags indicated.
131.00 € - 2345.00 €
Specifications
Accession Number | NP_033871.2 |
---|---|
Gene ID (Entrez) | 12043 |
Name | Bcl-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15917189
|
enQuireBio™
QP5710-EC-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957189
|
enQuireBio™
QP5710-EC-50UG |
50 μg |
324.00 €
50µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907189
|
enQuireBio™
QP5710-EC-100UG |
100 μg |
520.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937189
|
enQuireBio™
QP5710-EC-200UG |
200 μg |
835.00 €
200µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947189
|
enQuireBio™
QP5710-EC-500UG |
500 μg |
1440.00 €
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927189
|
enQuireBio™
QP5710-EC-1MG |
1 mg |
2345.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_033871.2 | |
Bcl-2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
12043 | |
Research Use Only | |
Bcl2 | |
Recombinant Protein | |
E. coli | |
GRTGYDNREI VMKYIHYKLS QRGYEWDAGD ADAAPLGAAP TPGIFSFQPE SNPMPAVHRD MAARTSPLRP LVATAGPALS PVPPVVHLTL RRAGDDFSRR YRRDFAEMSS QLHLTPFTAR GRFATVVEEL FRDGVNWGRI VAFFEFGGVM CVESVNREMS PLVDNIALWM TEYLNRHLHT WIQDNGGWDA FVELYGPSMR P | |
Greater than 90% as determined by SDS-PAGE. |