missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C5a Protein
A cDNA sequence encoding the C5a was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | C5a Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50 of Recombinant Mouse C5a as determined by its ability to induce N-acetyl-b-D-glucosaminidase release from differentiated U937 human histiocytic lymphoma cells was 5-20ng/ml. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994188
|
enQuireBio™
QP10541-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974188
|
enQuireBio™
QP10541-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984188
|
enQuireBio™
QP10541-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
C5a Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGLWGILCLL IFLDKTWGQE QTYVISAPKI LRVGSSENVV IQVHGYTEAF DATLSLKSYP DKKVTFSSGY VNLSPENKFQ NAALLTLQPN QVPREESPVS HVYLEVVSKH FSKSKKIPIT YNNGILFIHT DKPVYTPDQS VKIRVYSLGD DLKPAKRETV LTFIDPEGSE VDIVEENDYT GIISFPDFKI PSNPKYGVWT IKANYKKDFT TTGTAYFEIK EYVLPRFSVS IELERTFIGY KNFKNFEITV KARYFYNKVV PDAEVYAFFG LREDIKDEEK QMMHKATQAA KLVDGVAQIS FDSETAVKEL SYNSLEDLNN KYLYIAVTVT ESSGGFSEEA EIPGVKYVLS PYTLNLVATP LFVKPGIPFS IKAQVKDSLE QAVGGVPVTL MAQTVDVNQE TSDLETKRSI THDTDGVAVF VLNLPSNVTV LKFEIRTDDP ELPEENQASK EYEAVAYSSL SQSYIYIAWT ENYKPMLVGE YLNIMVTPKS PYIDKITHYN YLILSKGKIV QYGTREKLFS STYQNINIPV TQNMVPSARL LVYYIVTGEQ TAELVADAVW INIEEKCGNQ LQVHLSPDEY VYSPGQTVSL DMVTEADSWV ALSAVDRAVY KVQGNAKRAM QRVFQALDEK SDLGCGAGGG HDNADVFHLA GLTFLTNANA DDSHYRDDSC KEILRSKRNL HLLRQKIEEQ AAKYKHSVPK KCCYDGARVN FYETCEERVA RVTIGPLCIR AFNECCTIAN KIRKESPHKP VQLGRIHIKT LLPVMKADIR SYFPESWLWE IHRVPKRKQL QVTLPDSLTT WEIQGIGISD NGICVADTLK AKVFKEVFLE MNIPYSVVRG EQIQLKGTVY NYMTSGTKFC VKMSAVEGIC TSGSSAASLH TSRPSRCVFQ RIEGSSSHLV TFTLLPLEIG | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The ED50 of Recombinant Mouse C5a as determined by its ability to induce N-acetyl-b-D-glucosaminidase release from differentiated U937 human histiocytic lymphoma cells was 5-20ng/ml. | |
Mouse | |
Untagged | |
Mouse C5a was lyophilized from a concentrated (1 mg/ml) solution in 20mM PBS, pH 7.5 and 350mM NaCl. |