missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CALB2 Protein
A cDNA sequence encoding the CALB2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11259-2ug
Additional Details : Weight : 0.01000kg
Specifications
12308 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAGPQQQPPY LHLAELTASQ FLEIWKHFDA DGNGYIEGKE LENFFQELEK ARKGSGMMSK SDNFGEKMKE FMQKYDKNSD GKIEMAELAQ ILPTEENFLL CFRQHVGSSA EFMEAWRKYD TDRSGYIEAN ELKGFLSDLL KKANRPYDEP KLQEYTQTIL RMFDLNGDGK LGLSEMSRLL PVQENFLLKF QGMKLTSEEF NAIFTFYDKD GSGYIDENEL DALLKDLYEK NKKEMNIQQL TTYRKSVMSL AEAGKLYRKD LEIVLCSEPP V |
CALB2 Protein | |
Research Use Only | |
Calb2 | |
Mouse | |
Untagged | |
Calretinin protein at 100microg/ml in 50mM Tris-HCl, pH 7.5 and 10mM L-glutathione (reduced). |