missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD40L / CD154 / TNFSF5 Protein
Recombinant Protein
Brand: enQuireBio™ QP5411-5ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035746.2 | |
CD40L / CD154 / TNFSF5 Protein | |
Research Use Only | |
CD40LG | |
Recombinant Protein | |
E. coli | |
MQRGDEDPQI AAHVVSEANS NAASVLQWAK KGYYTMKSNL VMLENGKQLT VKREGLYYVY TQVTFCSNRE PSSQRPFIVG LWLKPSSGSE RILLKAANTH SSSQLCEQQS VHLGGVFELQ AGASVFVNVT EASQVIHRVG FSSFGLLKL | |
Greater than 95% as determined by SDS-PAGE. |
21947 | |
5 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Mouse CD40L / CD154 / TNFSF5 induced IL-8 production in human PBMCs detected via ELISA (typical ED50 is 1-10 ng/ml). Activity is also measured in HEK-Blue™ CD40L validation criteria in this assay is an ED50 of less than 50 ng/ml. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM arginine, pH7.5 |