missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD40L / CD154 / TNFSF5 Protein
Recombinant Protein
107.00 € - 4060.00 €
Specifications
Accession Number | NP_035746.2 |
---|---|
Gene ID (Entrez) | 21947 |
Name | CD40L / CD154 / TNFSF5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986069
|
enQuireBio™
QP5411-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966069
|
enQuireBio™
QP5411-25UG |
25 μg |
252.00 €
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946069
|
enQuireBio™
QP5411-100UG |
100 μg |
630.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976069
|
enQuireBio™
QP5411-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956069
|
enQuireBio™
QP5411-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_035746.2 | |
CD40L / CD154 / TNFSF5 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Mouse CD40L / CD154 / TNFSF5 induced IL-8 production in human PBMCs detected via ELISA (typical ED50 is 1-10 ng/ml). Activity is also measured in HEK-Blue™ CD40L validation criteria in this assay is an ED50 of less than 50 ng/ml. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM arginine, pH7.5 |
21947 | |
Research Use Only | |
CD40LG | |
Recombinant Protein | |
E. coli | |
MQRGDEDPQI AAHVVSEANS NAASVLQWAK KGYYTMKSNL VMLENGKQLT VKREGLYYVY TQVTFCSNRE PSSQRPFIVG LWLKPSSGSE RILLKAANTH SSSQLCEQQS VHLGGVFELQ AGASVFVNVT EASQVIHRVG FSSFGLLKL | |
Greater than 95% as determined by SDS-PAGE. |